Assay Design



Gene Transcripts

Gene Symbol
CHTOP
Entrez Gene ID
616638
Gene Name
chromatin target of PRMT1
Gene Aliases
C1orf77, C3H1orf77, FOP, SRAG
Chromosome Location
Chr.3: 16741096 - 16750610 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035481.2 NP_001030558.1 5 - 6 780 72
GenBank mRNA BC103347.1 - 5 - 6 781 72

View Full Product Details