Assay Design



Gene Transcripts

Gene Symbol
TSPAN3
Entrez Gene ID
616881
Gene Name
tetraspanin 3
Gene Aliases
-
Chromosome Location
Chr.21: 32177529 - 32204399 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035487.1 NP_001030564.1 2 - 3 379 91
GenBank mRNA BC102734.1 - 2 - 3 379 91

View Full Product Details