Assay Design



Gene Transcripts

Gene Symbol
SLC16A1
Entrez Gene ID
505775
Gene Name
solute carrier family 16 member 1
Gene Aliases
MCT1
Chromosome Location
Chr.3: 30380729 - 30411748 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037319.1 NP_001032396.1 2 - 3 416 106
GenBank mRNA AB250265.2 - 1 - 2 220 106
BC104598.2 - 2 - 3 416 106

View Full Product Details