Assay Design



Gene Transcripts

Gene Symbol
UBE2C
Entrez Gene ID
506962
Gene Name
ubiquitin conjugating enzyme E2 C
Gene Aliases
-
Chromosome Location
Chr.13: 74588078 - 74592792 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037449.2 NP_001032526.1 1 - 2 151 98
GenBank mRNA BC108195.1 - 1 - 2 152 98

View Full Product Details