Assay Design



Gene Transcripts

Gene Symbol
UBE2B
Entrez Gene ID
512207
Gene Name
ubiquitin conjugating enzyme E2 B
Gene Aliases
-
Chromosome Location
Chr.7: 45944974 - 45957592 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037459.2 NP_001032536.1 4 - 5 326 146
GenBank mRNA BC108202.1 - 4 - 5 327 146

View Full Product Details