Assay Design



Gene Transcripts

Gene Symbol
PCNP
Entrez Gene ID
615197
Gene Name
PEST proteolytic signal containing nuclear protein
Gene Aliases
-
Chromosome Location
Chr.1: 46002022 - 46016857 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038205.1 NP_001033294.1 2 - 3 292 128
GenBank mRNA BC108137.1 - 2 - 3 292 128

View Full Product Details