Assay Design



Gene Transcripts

Gene Symbol
LLPH
Entrez Gene ID
510518
Gene Name
LLP homolog, long-term synaptic facilitation factor
Gene Aliases
C12orf31
Chromosome Location
Chr.5: 47667169 - 47673189 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038522.1 NP_001033611.1 1 - 2 218 87
GenBank mRNA BC109741.1 - 1 - 2 218 87

View Full Product Details