Assay Design



Gene Transcripts

Gene Symbol
CD3G
Entrez Gene ID
281055
Gene Name
CD3g molecule
Gene Aliases
-
Chromosome Location
Chr.15: 29025582 - 29037075 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040472.2 NP_001035562.2 4 - 5 506 75
GenBank mRNA BC103010.1 - 4 - 5 502 75
DV802141.1 - 4 - 5 495 75
EH165860.1 - 4 - 5 507 75

View Full Product Details