Assay Design



Gene Transcripts

Gene Symbol
SCP2D1
Entrez Gene ID
505971
Gene Name
SCP2 sterol binding domain containing 1
Gene Aliases
C13H20orf79, C20orf79
Chromosome Location
Chr.13: 38704687 - 38706768 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040507.2 NP_001035597.1 1 - 1 648 141
GenBank mRNA BC109730.1 - 1 - 1 649 141

View Full Product Details