Assay Design



Gene Transcripts

Gene Symbol
TTC36
Entrez Gene ID
510592
Gene Name
tetratricopeptide repeat domain 36
Gene Aliases
-
Chromosome Location
Chr.15: 29194018 - 29198431 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040515.2 NP_001035605.1 1 - 2 176 76
GenBank mRNA BC102694.1 - 1 - 2 179 76

View Full Product Details