Assay Design



Gene Transcripts

Gene Symbol
COX20
Entrez Gene ID
615660
Gene Name
cytochrome c oxidase assembly factor COX20
Gene Aliases
FAM36A
Chromosome Location
Chr.16: 32421901 - 32429189 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040585.2 NP_001035675.1 3 - 4 339 82
GenBank mRNA BC109923.1 - 3 - 4 340 82

View Full Product Details