Assay Design



Gene Transcripts

Gene Symbol
ZNF33B
Entrez Gene ID
520684
Gene Name
zinc finger protein 33B
Gene Aliases
-
Chromosome Location
Chr.28: 12790859 - 12848389 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075810.1 NP_001069278.1 1 - 2 78 116
GenBank mRNA BC104558.1 - 1 - 2 78 116

View Full Product Details