Assay Design



Gene Transcripts

Gene Symbol
SULT1B1
Entrez Gene ID
521920
Gene Name
sulfotransferase family 1B member 1
Gene Aliases
ST1B1
Chromosome Location
Chr.6: 85190008 - 85233392 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075823.2 NP_001069291.1 6 - 7 898 70
GenBank mRNA BC102208.1 - 6 - 7 900 70

View Full Product Details