Assay Design



Gene Transcripts

Gene Symbol
ROPN1
Entrez Gene ID
527583
Gene Name
rhophilin associated tail protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 68349821 - 68384919 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075900.1 NP_001069368.1 1 - 2 246 83
GenBank mRNA BC102549.1 - 1 - 2 246 83

View Full Product Details