Assay Design



Gene Transcripts

Gene Symbol
DPH1
Entrez Gene ID
534099
Gene Name
diphthamide biosynthesis 1
Gene Aliases
DPH1-OVCA2
Chromosome Location
Chr.19: 23055699 - 23065910 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076899.2 NP_001070367.1 8 - 9 924 74
GenBank mRNA BC103408.1 - 8 - 9 925 74

View Full Product Details