Assay Design



Gene Transcripts

Gene Symbol
TMEM60
Entrez Gene ID
767911
Gene Name
transmembrane protein 60
Gene Aliases
-
Chromosome Location
Chr.4: 43450254 - 43454106 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076988.2 NP_001070456.1 1 - 2 180 131
GenBank mRNA BC102602.1 - 1 - 2 181 131

View Full Product Details