Assay Design



Gene Transcripts

Gene Symbol
AZI2
Entrez Gene ID
767932
Gene Name
5-azacytidine induced 2
Gene Aliases
-
Chromosome Location
Chr.22: 2757698 - 2798686 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077005.2 NP_001070473.1 3 - 4 730 127
GenBank mRNA BC103353.1 - 3 - 4 731 127

View Full Product Details