Assay Design



Gene Transcripts

Gene Symbol
PACRG
Entrez Gene ID
767959
Gene Name
parkin coregulated
Gene Aliases
-
Chromosome Location
Chr.9: 98192829 - 98258307 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077027.2 NP_001070495.1 2 - 3 520 82
GenBank mRNA BC108229.1 - 2 - 3 524 82

View Full Product Details