Assay Design



Gene Transcripts

Gene Symbol
MRPL23
Entrez Gene ID
508825
Gene Name
mitochondrial ribosomal protein L23
Gene Aliases
-
Chromosome Location
Chr.29: 49542745 - 49549286 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077884.1 NP_001071352.1 3 - 4 322 95
GenBank mRNA BC108192.1 - 3 - 4 322 95

View Full Product Details