Assay Design



Gene Transcripts

Gene Symbol
ZNF280B
Entrez Gene ID
517697
Gene Name
zinc finger protein 280B
Gene Aliases
SUHW2
Chromosome Location
Chr.17: 70979561 - 70990872 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077935.2 NP_001071403.2 1 - 2 132 125
GenBank mRNA BC102316.1 - 1 - 2 102 125

View Full Product Details