Assay Design



Gene Transcripts

Gene Symbol
VSTM2L
Entrez Gene ID
527903
Gene Name
V-set and transmembrane domain containing 2 like
Gene Aliases
C20orf102
Chromosome Location
Chr.13: 66799413 - 66838316 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077976.1 NP_001071444.1 2 - 3 504 65
GenBank mRNA BC105267.1 - 2 - 3 504 65

View Full Product Details