Assay Design



Gene Transcripts

Gene Symbol
IMP3
Entrez Gene ID
511560
Gene Name
IMP U3 small nucleolar ribonucleoprotein 3
Gene Aliases
BRMS2
Chromosome Location
Chr.21: 33254420 - 33255497 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079588.2 NP_001073056.1 1 - 1 550 78
GenBank mRNA BC102331.1 - 1 - 1 552 78

View Full Product Details