Assay Design



Gene Transcripts

Gene Symbol
LDAH
Entrez Gene ID
534992
Gene Name
lipid droplet associated hydrolase
Gene Aliases
C11H2orf43, C2orf43
Chromosome Location
Chr.11: 78103081 - 78191677 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079608.2 NP_001073076.1 4 - 5 526 81
GenBank mRNA BC102736.1 - 4 - 5 527 81

View Full Product Details