Assay Design



Gene Transcripts

Gene Symbol
JCHAIN
Entrez Gene ID
280821
Gene Name
joining chain of multimeric IgA and IgM
Gene Aliases
IGJ
Chromosome Location
Chr.6: 86032997 - 86042394 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175773.3 NP_786967.1 3 - 4 297 116
GenBank mRNA BC103426.1 - 3 - 4 300 116
L20311.1 - 3 - 4 270 116
U02301.1 - 3 - 4 270 116

View Full Product Details