Assay Design



Gene Transcripts

Gene Symbol
MBL1
Entrez Gene ID
497014
Gene Name
mannose binding lectin, liver (A)
Gene Aliases
MBL-A, MBLA
Chromosome Location
Chr.28: 35636034 - 35641256 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010994.3 NP_001010994.1 1 - 2 148 72
GenBank mRNA AB178774.1 - 1 - 2 68 72
BC109674.1 - 1 - 2 149 72

View Full Product Details