Assay Design



Gene Transcripts

Gene Symbol
CXCL13
Entrez Gene ID
511674
Gene Name
C-X-C motif chemokine ligand 13
Gene Aliases
-
Chromosome Location
Chr.6: 92284769 - 92291183 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015576.2 NP_001015576.1 3 - 4 314 69
GenBank mRNA AY911364.1 - 3 - 4 315 69
BC109998.1 - 3 - 4 314 69

View Full Product Details