Assay Design



Gene Transcripts

Gene Symbol
ARL14EP
Entrez Gene ID
523894
Gene Name
ADP ribosylation factor like GTPase 14 effector protein
Gene Aliases
ARF7EP, C11orf46, C15H11orf46
Chromosome Location
Chr.15: 61047589 - 61061505 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031761.3 NP_001026931.2 2 - 3 639 89
GenBank mRNA BC112569.1 - 2 - 3 642 89
BT020677.1 - 2 - 3 572 89

View Full Product Details