Assay Design



Gene Transcripts

Gene Symbol
PPP1R32
Entrez Gene ID
506337
Gene Name
protein phosphatase 1 regulatory subunit 32
Gene Aliases
C11orf66, C29H11orf66
Chromosome Location
Chr.29: 39953298 - 39962271 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038044.2 NP_001033133.1 5 - 6 806 62
GenBank mRNA BC111283.1 - 5 - 6 807 62

View Full Product Details