Assay Design



Gene Transcripts

Gene Symbol
C10H14orf93
Entrez Gene ID
506904
Gene Name
chromosome 10 C14orf93 homolog
Gene Aliases
-
Chromosome Location
Chr.10: 21839302 - 21858614 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038049.1 NP_001033138.1 1 - 2 129 52
GenBank mRNA BC111308.1 - 1 - 2 129 52

View Full Product Details