Assay Design



Gene Transcripts

Gene Symbol
GOT1L1
Entrez Gene ID
507913
Gene Name
glutamic-oxaloacetic transaminase 1 like 1
Gene Aliases
-
Chromosome Location
Chr.27: 33181298 - 33193163 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038058.1 NP_001033147.1 7 - 8 1035 61
GenBank mRNA BC111285.1 - 7 - 8 1035 61

View Full Product Details