Assay Design



Gene Transcripts

Gene Symbol
GTF2F2
Entrez Gene ID
509259
Gene Name
general transcription factor IIF subunit 2
Gene Aliases
-
Chromosome Location
Chr.12: 15282632 - 15418924 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038064.1 NP_001033153.1 6 - 7 551 86
GenBank mRNA BC111360.1 - 6 - 7 551 86

View Full Product Details