Assay Design



Gene Transcripts

Gene Symbol
ECRG4
Entrez Gene ID
515133
Gene Name
ECRG4 augurin precursor
Gene Aliases
C11H2orf40
Chromosome Location
Chr.11: 45829015 - 45838416 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038113.2 NP_001033202.1 2 - 3 228 66
GenBank mRNA BC110018.1 - 2 - 3 229 66

View Full Product Details