Assay Design



Gene Transcripts

Gene Symbol
TRAPPC6B
Entrez Gene ID
521470
Gene Name
trafficking protein particle complex 6B
Gene Aliases
-
Chromosome Location
Chr.21: 49145029 - 49156067 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038134.1 NP_001033223.1 4 - 5 494 85
GenBank mRNA BC109723.1 - 4 - 5 494 85

View Full Product Details