Assay Design



Gene Transcripts

Gene Symbol
REEP6
Entrez Gene ID
522480
Gene Name
receptor accessory protein 6
Gene Aliases
-
Chromosome Location
Chr.7: 43895971 - 43901428 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038135.2 NP_001033224.1 4 - 5 802 64
GenBank mRNA BC109592.1 - 4 - 5 803 64

View Full Product Details