Assay Design



Gene Transcripts

Gene Symbol
TMEM30C
Entrez Gene ID
531379
Gene Name
transmembrane protein 30C
Gene Aliases
-
Chromosome Location
Chr.1: 44483383 - 44508874 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038150.2 NP_001033239.1 6 - 7 702 107
GenBank mRNA BC111328.1 - 6 - 7 703 107

View Full Product Details