Assay Design



Gene Transcripts

Gene Symbol
SPATA22
Entrez Gene ID
539692
Gene Name
spermatogenesis associated 22
Gene Aliases
-
Chromosome Location
Chr.19: 24197779 - 24212672 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038186.2 NP_001033275.1 4 - 5 376 136
GenBank mRNA BC111158.1 - 4 - 5 377 136

View Full Product Details