Assay Design



Gene Transcripts

Gene Symbol
SPATA22
Entrez Gene ID
539692
Gene Name
spermatogenesis associated 22
Gene Aliases
-
Chromosome Location
Chr.19: 24197779 - 24212672 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038186.2 NP_001033275.1 8 - 9 1057 114
GenBank mRNA BC111158.1 - 8 - 9 1058 114

View Full Product Details