Assay Design



Gene Transcripts

Gene Symbol
ILF2
Entrez Gene ID
539760
Gene Name
interleukin enhancer binding factor 2
Gene Aliases
-
Chromosome Location
Chr.3: 16711945 - 16722477 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038187.2 NP_001033276.1 5 - 6 364 83
GenBank mRNA BC111202.1 - 5 - 6 365 83

View Full Product Details