Assay Design



Gene Transcripts

Gene Symbol
SPATA6L
Entrez Gene ID
613587
Gene Name
spermatogenesis associated 6 like
Gene Aliases
C8H9orf68
Chromosome Location
Chr.8: 39858145 - 39959303 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205485.1 NP_001192414.1 7 - 8 829 104
GenBank mRNA BC111296.1 - 11 - 12 1342 104

View Full Product Details