Assay Design
Gene Transcripts
- Gene Symbol
- SPATA6L
- Entrez Gene ID
-
613587
- Gene Name
- spermatogenesis associated 6 like
- Gene Aliases
-
C8H9orf68
- Chromosome Location
-
Chr.8: 39858145 - 39959303 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC111296.1 |
- |
5 - 6 |
515 |
74 |
View Full Product Details