Assay Design
Gene Transcripts
- Gene Symbol
- AZIN2
- Entrez Gene ID
-
506961
- Gene Name
- antizyme inhibitor 2
- Gene Aliases
-
ADC
- Chromosome Location
-
Chr.2: 120568183 - 120614958 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC109524.1 |
- |
13 - 14 |
1931 |
59 |
View Full Product Details