Assay Design



Gene Transcripts

Gene Symbol
AZIN2
Entrez Gene ID
506961
Gene Name
antizyme inhibitor 2
Gene Aliases
ADC
Chromosome Location
Chr.2: 120568183 - 120614958 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038510.3 NP_001033599.2 8 - 9 1142 97
GenBank mRNA BC109524.1 - 8 - 9 1143 97

View Full Product Details