Assay Design



Gene Transcripts

Gene Symbol
CXHXorf21
Entrez Gene ID
513911
Gene Name
chromosome X CXorf21 homolog
Gene Aliases
C9HXorf21, CXorf21
Chromosome Location
Chr.X: 113007471 - 113020233 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038537.2 NP_001033626.1 1 - 2 165 65
GenBank mRNA BC109631.1 - 1 - 2 167 65

View Full Product Details