Assay Design



Gene Transcripts

Gene Symbol
MMP23
Entrez Gene ID
527590
Gene Name
matrix metallopeptidase 23
Gene Aliases
MMP23B
Chromosome Location
Chr.16: 51003855 - 51006360 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038556.2 NP_001033645.1 2 - 3 308 66
GenBank mRNA BC109902.1 - 2 - 3 310 66

View Full Product Details