Assay Design



Gene Transcripts

Gene Symbol
ARL3
Entrez Gene ID
540040
Gene Name
ADP ribosylation factor like GTPase 3
Gene Aliases
-
Chromosome Location
Chr.26: 23292986 - 23322870 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038567.2 NP_001033656.1 3 - 4 296 112
GenBank mRNA BC109565.1 - 3 - 4 297 112

View Full Product Details