Assay Design



Gene Transcripts

Gene Symbol
SRPX
Entrez Gene ID
337918
Gene Name
sushi repeat containing protein X-linked
Gene Aliases
ext1
Chromosome Location
Chr.X: 105052751 - 105194337 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040489.1 NP_001035579.1 6 - 7 866 81
GenBank mRNA AJ505581.1 - 1 - 2 69 81
BC109914.1 - 6 - 7 866 81

View Full Product Details