Assay Design



Gene Transcripts

Gene Symbol
TM2D3
Entrez Gene ID
512588
Gene Name
TM2 domain containing 3
Gene Aliases
-
Chromosome Location
Chr.21: 28805944 - 28850666 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040518.2 NP_001035608.1 1 - 2 104 84
GenBank mRNA BC109864.1 - 1 - 2 105 84

View Full Product Details