Assay Design



Gene Transcripts

Gene Symbol
RPL7A
Entrez Gene ID
513128
Gene Name
ribosomal protein L7a
Gene Aliases
-
Chromosome Location
Chr.11: 104255845 - 104259161 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040520.2 NP_001035610.1 1 - 2 30 59
GenBank mRNA BC109810.1 - 1 - 2 31 59

View Full Product Details