Assay Design



Gene Transcripts

Gene Symbol
DYDC1
Entrez Gene ID
613509
Gene Name
DPY30 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.28: 35707659 - 35745292 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040571.2 NP_001035661.1 7 - 8 735 123
GenBank mRNA BC109581.1 - 7 - 8 736 123

View Full Product Details