Assay Design



Gene Transcripts

Gene Symbol
APOA2
Entrez Gene ID
505394
Gene Name
apolipoprotein A2
Gene Aliases
-
Chromosome Location
Chr.3: 8282869 - 8284124 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045916.2 NP_001039381.1 1 - 2 39 68
GenBank mRNA BC111607.1 - 1 - 2 41 68

View Full Product Details